Learn More
PGI™ FireLine™ Wildland Jumpsuits
Maximum ease of movement with minimum bulk.
Manufacturer:  PGISupplier Diversity Partner 6501872MR
 View more versions of this product
Catalog No. 19-120-3113
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
NFPA 1977
Includes: Radio pocket, microphone holder, cell phone pocket, D-ring loop, CAL OSHA label
38 to 40 in.
Alpine
Medium
14.7 to 16.5cm
31 to 34 in.
Regular
Zipper
Indura™ Ultra soft™ Fabric
Collared
Yellow
Designed to allow maximum ease of movement with minimum bulk
5.8 to 6.5 in.
78.7 to 86.3cm
Back elastic