Learn More
PGI PGI™ Cobra™ Firefighting Hoods
Feature flat-stitched seams for improved comfort and fit
Manufacturer:  PGI 3049298
 View more versions of this product
Catalog No. 19-816-683
Add to cart
Description
Description
Hoods are available in two flame-resistant materials. Carbon Shield hood is a unique blend of high-temperature-resistant carbon and high-strength aramid that is lightweight and extremely comfortable to wear. Nonflammable in extreme heat, this melt-proof, nonstiffening fabric will glow but not burn. DuPont™ Nomex™ and Lenzing Fiber's Lenzing FR™ blended together to create exceptionally durable, comfortable, flame-resistant fabric, Comfort-Plus™, for hood linings.
Common Features
  • 0.5 in. wide elastic surrounds face opening and stretches to full 17 in. for easy donning and snug fit around SCBA mask
  • Bottom edges are bound with self-material bias binding, 100% Nomex thread
  • Rib-knit construction offers better stretch and recovery
Classic™ Style Hoods
  • Provide double layer coverage of the neck, upper chest and top of shoulder blades
  • Contoured shoulder blades reduce bunching of bib
Ultimate™ Style Hood
  • Designed for maximum protection
  • Covers a greater percentage of the chest, shoulders and shoulder blades
Compliance
Meet OSHA 29 CFR 1910.269 and CAL-OSHA Title 8 Article 10.0, Sections 3406 and 3410(d). UL classified to NFPA™ 1971, 2007 Edition.
Specifications
Specifications
OSHA 29 CFR 1910.269, CAL-OSHA, UL, NFPA 1971
Carbon Shield™
X-heavy 0.5 in. W elasticized face opening, cover stitched bound drape
Flat stitched
Black
Each
Carbon Shield™