PGI™ Cobra™ Firefighting Hoods
Feature flat-stitched seams for improved comfort and fit
Manufacturer: PGISupplier Diversity Partner 3018185
 View more versions of this product
Catalog No. 19-500-373
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance OSHA 29 CFR 1910.269, CAL-OSHA, UL, NFPA 1971
    Hood Material 2-ply
    Quantity Each
    Includes Comfort Plus liner for cooler, drier wear, X-heavy 0.5 in. W elasticized face opening, cover stitched bound drape, notched shoulders
    Material Nomex™, Lenzing FR™
    Seam Type Flat stitched
    Color White
  • Description & Specifications
Related Products
Product Content Feedback