Learn More
PGLYRP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 180821AP
Catalog No. 50-173-1975
Add to cart
Description
Description
Primary immune recognition is based on structures common among invading pathogens. Bacterial surface molecules, such as lipopolysaccharide (LPS) and peptidoglycan (PGN), are known to elicit immune reactions ranging from cytokine release to fever. Recently, a family of proteins called peptidoglycan recognition protein (PGRP) has been identified in mouse and human that binds to peptidoglycans expressed on Gram-positive bacteria. Peptidoglycan (PGN) is an essential cell wall component of virtually all bacteria (1, 2) and, thus, it is an excellent target for recognition by the eukaryotic innate immune system. The PGRPs (PGRP-L, PGRP-S, PGRP-Ia, and PGRP-I beta) define a new family of human pattern recognition molecules (3). PGRP-L is primarily expressed in the liver. Although liver is not considered a primary immune organ, liver participates in host defenses by producing acute phase proteins (by hepatocytes) in response to infections and by clearing microorganisms from blood (4). PGRP-1a mRNA is highly expressed in esophagus, tonsils and thymus and to lesser extent in the stomach, rectum, and brain.
Specifications
Specifications
PGLYRP3
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A1A547, Q96LB9
PGLYRP3, PGLYRPIalpha, PGRP I alpha, PGRP Ialpha, PGRPIA
Rabbit
IgG
150 μL
-20°C
Primary
114771, 242100, 499658
Western Blot
Unconjugated
PGLYRP3
Liquid
Pglyrp3
PGLYRP3 Fusion Protein Ag12792
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat