Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGM3 Mouse anti-Human, Mouse, Clone: OTI1A11, liquid, TrueMAB™
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA504859

Encompass_Preferred

Catalog No. 50-167-5459


Add to Cart

Description

Description

Catalyzes the conversion of GlcNAc-6-P into GlcNAc-1-P during the synthesis of uridine diphosphate/UDP-GlcNAc, a sugar nucleotide critical to multiple glycosylation pathways including protein N- and O-glycosylation.
Specifications

Specifications

PGM3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O95394, Q9CYR6
Pgm3
Full length human recombit protein of human PGM3 produced in HKE293T cell.
100 μL
Primary
Human, Mouse
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI1A11
Unconjugated
PGM3
AGM1, IMD23, PAGM, PGM 3
Mouse
Affinity Chromatography
RUO
109785, 5238
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit