Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGP9.5 175-191 Guinea Pig anti-Human, Mouse, Rat, Polyclonal, Neuromics™

Guinea Pig Polyclonal Antibody

Manufacturer:  Neuromics, IncSupplier Diversity Partner GP14104150UL

Encompass

 View more versions of this product

Catalog No. 50-210-0009


Add to cart

Description

Description

PGP9.5 175-191 Polyclonal antibody specifically detects PGP9.5 175-191 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry
Specifications

Specifications

PGP9.5 175-191
Polyclonal
Unconjugated
Uchl1
Liquid
Guinea Pig
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry
Conc. Not Determined
whole serum with 0.02% sodium azide
P09936, Q00981, Q9R0P9
AW822034; BOS_6576; C88048; epididymis luminal protein 117; epididymis secretory protein Li 53; gad; gracile axonal dystrophy; HEL-117; HEL-S-53; LOC100357806; NDGOA; Neuron cytoplasmic protein 9.5; OTTHUMP00000218137; OTTHUMP00000218139; OTTHUMP00000218140; OTTHUMP00000218141; PARK5; PGP 9.5; PGP9.5; PGP95; protein gene product 9.5; R75593; ubiquitin carboxyl-terminal esterase L1; ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase); Ubiquitin carboxyl-terminal hydrolase isozyme L1; ubiquitin carboxyl-terminal hydrolase L1; ubiquitin carboxy-terminal hydrolase L1; ubiquitin C-terminal hydrolase; ubiquitin C-terminal hydrolase L1; ubiquitin C-terminal hydrolase-L1; Ubiquitin thioesterase L1; ubiquitin thiolesterase; ubiquitin thiolesterase L1; UCHL; UCHL1; UCH-L1; UCHL-1
GASSEDTLLKDAAKVCR
150 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
22223, 29545, 7345
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit