Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGP9.5 Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, Neuromics™ Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

$213.28 - $476.16

Specifications

Antigen PGP9.5
Concentration Conc. Not Determined
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-0034 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
RA12103150UL
150 μL
Each for $476.16
Add to cart
 
50-210-0035 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
RA1210350UL
50 μL
Each for $213.28
Add to cart
 
Description

Description

PGP9.5 Polyclonal antibody specifically detects PGP9.5 in Human, Mouse, Porcine, Rat samples. It is validated for Immunohistochemistry, Western Blot
Specifications

Specifications

PGP9.5
Immunohistochemistry, Western Blot
Unconjugated
Rabbit
Human, Mouse, Porcine, Rat
P09936, Q00981, Q6SEG5, Q9R0P9
22223, 29545, 396637, 7345
IgG
Primary
Antibody
Conc. Not Determined
Polyclonal
Liquid
RUO
whole serum with 0.05% sodium azide
AW822034; BOS_6576; C88048; epididymis luminal protein 117; epididymis secretory protein Li 53; gad; gracile axonal dystrophy; HEL-117; HEL-S-53; LOC100357806; NDGOA; Neuron cytoplasmic protein 9.5; OTTHUMP00000218137; OTTHUMP00000218139; OTTHUMP00000218140; OTTHUMP00000218141; PARK5; PGP 9.5; PGP9.5; PGP95; protein gene product 9.5; R75593; ubiquitin carboxyl-terminal esterase L1; ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase); Ubiquitin carboxyl-terminal hydrolase isozyme L1; ubiquitin carboxyl-terminal hydrolase L1; ubiquitin carboxy-terminal hydrolase L1; ubiquitin C-terminal hydrolase; ubiquitin C-terminal hydrolase L1; ubiquitin C-terminal hydrolase-L1; Ubiquitin thioesterase L1; ubiquitin thiolesterase; ubiquitin thiolesterase L1; UCHL; UCHL1; UCH-L1; UCHL-1
ASSEDTLLKDAAKVCR
Polyclonal
-20° C, Avoid Freeze/Thaw Cycles
Uchl1
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit