Learn More
PGPEP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 195661AP
Catalog No. 50-173-2459
Add to cart
Description
Description
Pyroglutamyl peptidase I (EC 3.4.19.3) catalyzes the hydrolysis of N-terminal pyroglutamyl residues from oligopeptides and proteins.
Specifications
Specifications
PGPEP1
0.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q76IC5, Q9ESW8, Q9NXJ5
5 oxoprolyl peptidase, PAP I, Pcp, PGP, PGP I, PGPEP1, PGPI, Pyroglutamyl aminopeptidase I, Pyroglutamyl peptidase 1, pyroglutamyl peptidase I
Rabbit
IgG
150 μL
-20°C
Primary
290648, 54858, 66522
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
PGPEP1
Liquid
Pgpep1
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat