Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGRMC1 Mouse anti-Human, Clone: 4A68, MilliporeSigma™
Mouse Monoclonal Antibody
$177.00 - $450.00
Specifications
Antigen | PGRMC1 |
---|---|
Clone | 4A68 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF265325
|
MilliporeSigma
MABF265325UL |
25 μL |
Each for $177.00
|
|
|||||
MABF2653100
|
MilliporeSigma
MABF2653100UL |
100 μL |
Each for $450.00
|
|
|||||
Description
PGRMC1 Monoclonal antibody specifically detects PGRMC1 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Western Blotting.Specifications
PGRMC1 | |
0.5 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
IgG1 κ | |
Protein G purified |
4A68 | |
Flow Cytometry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
H9 cells cultured with retinoic acid | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title