Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGRMC1 Rabbit anti-Human, DyLight 488, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297696G
Description
PGRMC1 Polyclonal specifically detects PGRMC1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PGRMC1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
HPR6.6PGRMC, membrane-associated progesterone receptor component 1, MPR, progesterone binding protein, progesterone receptor membrane component 1 | |
Produced in rabbits immunized with E. coli-derived Human PGRMC1 fragment. | |
0.1 mL | |
Cancer | |
10857 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title