pH 10 Reference Standard solution Buffer, Color coded blue, Macron Fine Chemicals™

macron

Manufacturer: Macron 009910

Packaging Poly
Quantity 4L

 View more versions of this product

Catalog No. 14-650-404

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Liquid
    Chemical Name or Material Buffer, pH 10 Reference standard solution

    Specifications

    pH 9.99 to 10.01 (5% soln. at 25°C)
    Color Blue
    Physical Form Liquid
    Quantity 4L
    Appearance Clear, blue solution
    Packaging Poly
    Traceability to NIST Yes
    Chemical Name or Material Buffer, pH 10 Reference standard solution

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit