Whatman™ pH Indicator Papers, CF Strips: PROMO OFFER

Manufacturer: Whatman™ 2614991PM

 View more versions of this product

Catalog No. 09-876-18PM

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    For Use With (Application) For the rapid determination of pH values has led to many applications in laboratories and industry
    Graduations 0.5 pH units
    Includes Plastic supports, paper segments on each strip, complementary indicator charts
    Quantity 100 Pack
    Product Type pH Indicator and CF Test Strip
    pH Range 4.5 to 10

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit