Learn More
Enzo Life Sciences Phaclofen (25mg). CAS: 114012-12-3
SDP
Supplier:  Enzo Life Sciences BMLEA1270025
Encompass_Preferred
Selective GABAB receptor antagonist. Antagonizes the postsynaptic action of baclofen (BML-EA128). Alternative name: (+/-)-3-Amino-2-(4-chlorophenyl)-propylphosphomic acid. Purity: ≥98% (HPLC). Solubility: Soluble in water, methanol or DMSO. Long Term Storage: Ambient.
Catalog No. 50-201-0533
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.