Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phaclofen, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

114012-12-3
C9H13ClNO3P
249.631
VSGNGLJPOGUDON-UHFFFAOYSA-N
(RS)-3-Amino-2-(4-chlorophenyl)propylphosphonic acid
1641

Weak, selective GABAB antagonist

Selective GABAB antagonist. Phosphono analog of GABAB agonist baclofen (Cat. No. 0417).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
SMILES C1=CC(=CC=C1C(CN)CP(=O)(O)O)Cl
IUPAC Name [3-amino-2-(4-chlorophenyl)propyl]phosphonic acid
Formula Weight 249.63
Chemical Name or Material Phaclofen
Grade >97%

Recommended Storage : Store at Room Temperature