Learn More
Thermo Scientific™ Phadia™ β 2-Glycoprotein I Antibodies for Antiphospholipid Syndrome
Intended for the in vitro quantitative measurement of antibodies directed to β2-Glycoprotein I in human serum and plasma to aid in the diagnosis of antiphospholipid syndrome (APS) and to evaluate the thrombotic risk in patients with systemic lupus erythematosus (SLE).
Manufacturer:  Thermo Scientific™ 14553101
 View more versions of this product
Catalog No. 14-553-101P
Add to cart
This item is not returnable. View return policy
Description
Description
If present in the patient's specimen, antibodies to β2-Glycoprotein I bind to their specific antigen. After washing away non-bound antibodies, enzyme-labeled antibodies against human IgG (IgM, IgA) antibodies (EliA IgG, IgM, IgA Conjugate™) are added to form an antibody-conjugate complex. After incubation, non-bound conjugate is washed away and the bound complex is incubated with a Development Solution. After stopping the reaction, the fluorescence in the reaction mixture is measured. To evaluate test results, the response for patient samples is compared directly to the response for calibrators.
  • The EliA 2-Glycoprotein I IgG (IgM, IgA) Wells are coated with human β2-Glycoprotein I antigen
  • The higher the responseβ value, the more specific IgG (IgM, IgA) is present in the specimen
  • All conjugates are isotype-specific -- mixed conjugate is not available
Specifications
Specifications
EliA™ Beta2-Glycoprotein I IgA
In-vitro quantitative measurement of Antibodies directed to beta2-Glycoprotein I in Human Serum and Plasma to aid in the diagnosis of antiphospholipid syndrome
2 x 12 Tests