Learn More
Thermo Scientific™ Phadia™ Controls for ImmunoCAP™ Celiac Disease Autoimmunity Tests
ImmunoCAP Gliadin IgA/IgG quantitatively measures IgA and IgG specific antibodies or gliadin in human serum or plasma
Manufacturer:  Thermo Scientific™ 10948601
 View more versions of this product
Catalog No. 10-948-601P
Add to cart
Description
Description
Measurement of circulating anti-gliadin antibodies can be used as an aid in the clinical diagnosis of patients with celiac disease and dermatitis herpetiformis, in conjunction with other clinical findings.