Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
BioVision IncSupplier Diversity Partner EZCell™ Phagocytosis Assay Kit (Green Zymosan): EZCellTM Phagocytosis Assay Kit (Green Zymosan): Non-radioactive, high-throughput assay quantify Phagocytosis or screening Phagocytosis effectors. 100 Assays DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K397100
EZCell™ Phagocytosis Assay Kit (Green Zymosan): Phagocytosis in mammals serves as an important first line defense mechanism against invading pathogens. It is also essential for continuous clearance of dying cells, tissue remodeling, and acquisition of nutrients for some cells. Phagocytosis is a specific form of endocytosis initiated by recognition and binding of foreign particles by cell surface receptors, followed by their engulfment, and formation of phagosomes. Zymosan prepared from yeast cell wall (Saccharomyces cerevisiae), and consisting of protein-carbohydrate complexes is frequently used as a pathogen in phagocytosis assays. BioVision's EZCellTM Phagocytosis Assay Kit (Green Zymosan) utilizes pre-labeled Zymosan particles as a tool for rapid and accurate detection and quantification of in vitro phagocytosis by fluorescent microscope, spectrophotometer or flow cytometry. . BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9735