Learn More
Cell Biolabs IncSupplier Diversity Partner PHAGOCYTOSIS KT 96W ZYMOSAN  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner CBA224
CytoSelect 96-Well Phagocytosis Assay, Zymosan Substrate provides a more accurate, high-throughput alternative to the standard phagocytosis assay. May be adapted for use with 24-well or 48-well plates.
Catalog No. 50-672-673