Learn More
Nor-Lake™ Scientific Pharmacy Standard Economy Solid Door Refrigerator, Bottom Mount
Forced air directional refrigeration system that provide superior temperature uniformity and recovery after door openings.
$3935.00 - $7811.00
Specifications
Defrost Cycle
Casters Yes
Amperage 10 A
Display Digital
Ports 1
View More Specs
Products 7
Catalog Number Mfr. No. Capacity (English) Refrigerator Door Style Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Refrigerator Door Style Price Quantity & Availability  
22-651-225
View Documents
Nor-Lake™ Scientific
PHER121WWW/0
12 cu. ft. Solid Swing
Each for $3,935.00
Add to cart
22-651-226
View Documents
Nor-Lake™ Scientific
PHER161WWW/0
16 cu. ft. Solid Swing
Each for $4,005.00
Add to cart
22-651-227
View Documents
Nor-Lake™ Scientific
PHER231WWW/0
23 cu. ft. Solid Swing
Each for $4,710.00
Add to cart
22-651-228
View Documents
Nor-Lake™ Scientific
PHER261WWW/0
26 cu. ft. Solid Swing
Each for $4,767.00
Add to cart
22-651-229
View Documents
Nor-Lake™ Scientific
PHER362WWW/0
36 cu. ft. Solid Swing
Each for $6,022.00
Add to cart
22-651-230
View Documents
Nor-Lake™ Scientific
PHER492WWW/0
49 cu. ft. Solid Swing
Each for $6,528.00
Add to cart
22-651-231
View Documents
Nor-Lake™ Scientific
PHER723WWW/0
72 cu. ft. Solid Swing
Each for $7,811.00
Add to cart
Description
Description
  • Microprocessor temperature controller with audible and visual alarms
  • Digital temperature display
  • Freestanding thermometer with 3 year certificate of calibration
  • 2 probes; 1 in air and 1 in sample bottle
  • Remote alarm contacts
  • Keyed door locks, leveling legs, and pharmacy refrigerator/freezer toolkit, temperature logs and CDC approved power cord labels
Specifications
Specifications
Cycle
10 A
1
NEMA 5-15 P
60 Hz
R134a
115 V
Yes
Digital
1 ph
High density urethane foam
Audible and visual high and low temperature alarms
Microprocessor