Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Pharmalyte™ Carrier Ampholytes

Use in Isoelectric focusing. Cytiva Pharmalyte™ Carrier Ampholytes prepared by the co-polymerization of glycine, glycylglycine, amines, and epichlorohydrin, are available in five broad-range and four narrow-range pH intervals.

$1063.00 - $1443.00

Specifications

Color Colorless to Light Yellow
For Use With (Equipment) PhastSystem Automated Electrophoresis Unit
Product Type Pharmalyte carrier Ampholyte
Form Liquid
Formulation Co-polymerization of Glycine, Glycylglycine, Amines and Epichlorohydrin
View More Specs

Products 5
Catalog Number Mfr. No. For Use With (Application) pH Range Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) pH Range Price Quantity & Availability  
45-000-076
View Documents
Cytiva
17045301
IEF 5 to 8
Each for $1,422.00
Add to Cart
 
09-928-075
View Documents
Cytiva
17045201
IEF 4 to 6.5
Each for $1,349.00
Add to Cart
 
45-000-074
View Documents
Cytiva
17045101
IEF 2.5 to 5
Each for $1,443.00
Add to Cart
 
45-000-077
View Documents
Cytiva
17045501
IEF 8 to 10.5
Each for $1,354.00
Add to Cart
 
45-000-078
View Documents
Cytiva
17045601
IEF 3 to 10
Each for $1,063.00
Add to Cart
 
Description

Description

  • Co-polymerisates of glycine, glycylglycine, amines and epichlorhydrin
  • Form extremely stable, linear pH gradients under the influence of electric fields
  • Available in both narrow and broad pH ranges to suit specific application
  • Broad ranges can be used directly for agarose, Ultrodex or polyacrylamide gels
  • Narrow ranges provide high resolution for dedicated applications using polyacrylamide gels

Specifications

Specifications

Colorless to Light Yellow
Pharmalyte carrier Ampholyte
Co-polymerization of Glycine, Glycylglycine, Amines and Epichlorohydrin
25mL
PhastSystem Automated Electrophoresis Unit
Liquid
IEF
2° to 8°C
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit