Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Pharmalyte™ Carrier Ampholytes PROMO

Cytiva Pharmalyte™ Carrier Ampholytes PROMO for use in isoelectric focusing

Specifications

Color Colorless to Light Yellow
Format Liquid
Formulation Co-polymerization of Glycine, Glycylglycine, Amines and Epichlorohydrin
Quantity 25 mL
Storage Requirements 2°C to 8°C
View More Specs

Products 3
Catalog Number Mfr. No. pHRange Price Quantity & Availability  
Catalog Number Mfr. No. pHRange Price Quantity & Availability  
45-000-076LR0
View Documents
Cytiva
29489318
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
45-000-077LR0
View Documents
Cytiva
29489317
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
45-000-078LR0
View Documents
Cytiva
29489316
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

  • Co-polymerisates of glycine, glycylglycine, amines and epichlorhydrin
  • Form extremely stable, linear pH gradients under the influence of electric fields
  • Available in both narrow and broad pH ranges to suit specific application
  • Broad ranges can be used directly for agarose, Ultrodex or polyacrylamide gels
  • Narrow ranges provide high resolution for dedicated applications using polyacrylamide gels

Specifications

Specifications

Colorless to Light Yellow
Co-polymerization of Glycine, Glycylglycine, Amines and Epichlorohydrin
2°C to 8°C
Liquid
25 mL
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit