Learn More
Cytiva (Formerly GE Healthcare Life Sciences) PhastGel™ Precast IEF Gels
Precast homogeneous gels containing Pharmalyte as carrier ampholyte to generate stable, linear pH gradients with uniform conductivity across the entire pH range and allow high field strengths for high resolution separations.
$222.00 - $288.00
Specifications
Separation of Protein
Separation Type Isoelectric Focusing
Gel Type Pharmalyte
Gel Thickness 0.45 mm
View More Specs
Products 3
Catalog Number Mfr. No. pH Range Price Quantity & Availability  
Catalog Number Mfr. No. pH Range Price Quantity & Availability  
45-000-098
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
17054501
pH 5 to 8
Pack of 10 for $288.00
Add to cart
45-000-097
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
17054401
pH 4 to 6.5
Pack of 10 for $222.00
Add to cart
45-000-096
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
17054301
pH 3 to 9
Pack of 10 for $226.50
Add to cart
Description
Description
  • PhastGel IEF gels
  • Homogeneous gels in various pH ranges for separations of proteins by isoelectric point
Specifications
Specifications
Protein
Isoelectric Focusing
Pharmalyte
0.45 mm