Learn More
GE Healthcare PhastGel™ Precast IEF Gels
Precast homogeneous gels containing Pharmalyte as carrier ampholyte to generate stable, linear pH gradients with uniform conductivity across the entire pH range and allow high field strengths for high resolution separations.
Manufacturer:  GE Healthcare 17054501
 View more versions of this product
Catalog No. 45-000-098
Add to cart
Description
Description
  • PhastGel IEF gels
  • Homogeneous gels in various pH ranges for separations of proteins by isoelectric point
Specifications
Specifications
125°C
5% T, 3% C
Pharmalyte
Isoelectric Focusing
PhastGel IEF – 5-8
Containing 2 to 6% Pharmalyte as carrier Ampholyte
4.8 cm
82° to 86°C
pH 5 to 8
0.017 in.
5 cm
GelBond polyester film
10 Pack
0.45 mm
pH 5 to 8
Protein
Used for charecterizing and analysing mixtures of proteins with smaller pI differentials
1.88 in.
4.8 cm
71.08
4 - 8°C
1.9 in.
Precast Gel