Learn More
GE Healthcare PhastGel™ Precast Gels
Select from homogeneous and gradient polyacrylamide gels. GE Healthcare PhastGel™ Precast Gels save time and improve reproducibility.
Manufacturer:  GE Healthcare 17067701
 View more versions of this product
Catalog No. 45-000-127
Add to cart
Description
Description
  • Select from homogeneous and gradient polyacrylamide gels
  • Separate proteins and peptides on denaturing and native
  • Save time and improve reproducibility using precast gels with native or denaturing buffer strips and staining kits, buffer strips, and staining kits
  • Achieve detection sensitivity comparable to that of radioisotopes with ready-to-use silver staining kits
  • Gels are cast on a plastic backing for ease of handling
PhastGel homogenous gels
  • Available in 7.5%, 12.5%, and 20% polyacrylamide with stacking gel, for flexibility in separation, extending the resolution between proteins within a narrow MW range
  • A high density PhastGel (20%) is also available containing PEG to optimise separations below Mr 20000 increasing resolution
PhastGel homogenous gels
  • Available in concentration gradients of 4-15%,10-15% and 8-25% with stacking gel, for separation of complex mixtures, giving a good overall view of total composition within a sample
Specifications
Specifications
10 Pack
5%
Pharmalyte
Denaturing, Native
PhastGel – 5.0
Containing 2 to 6% Pharmalyte as carrier Ampholyte
4.3 cm
- 20 to 80°C
1.9 in.
Precast Gel
PhastGel Cassette, Rehydration Cassette
0.45 mm
10 to 50 Mr
Protein
Used for industrial applications such as analytical chemistry and research
1.69 in.
4.3 cm
0.017 in.
5 cm
Polyester film