Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHD3, Polyclonal, Bethyl Laboratories Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Bethyl Laboratories, IncSupplier Diversity Partner A300327AM

 View more versions of this product

Catalog No. 50-155-7381


Add to cart

Description

Description

The recommended shelf life for this product is 1 year from date of receipt.

HIF prolyl hydroxylase 3 is a prolyl hydroxylase that modifies HIF-alpha. Classic prolyl hydroxylases are found in the endoplasmic reticulum and modify collagen, whereas HIF is an intracellular protein and the HPH sites do not resemble those modifying collagen. HIF is a transcriptional complex that plays a critical role in oxygen homeostasis. HPH is an essential component of the pathway through which cells sense oxygen. In the presence of oxygen, HPHs convert specific prolyl residues in HIF-alpha to hydroxyproline, leading to HIF-alpha destruction. Low oxygen levels, sensed at the cellular level, cause the HIF conversion to be reduced so that HIF is stable and there is increased angiogenesis. HIF prolyl hydroxylase 1, specifically, catalyzes the posttranslational formation of 4-hydroxyproline in HIF alpha proteins. It hydroxylates HIF-1 alpha at Pro(564) and HIF-2 alpha. It targets HIF through the hydroxylation for proteasomal degradation via the von Hippel-Lindau ubiquitylation complex. It may also play a role in cell growth regulation in muscle cells and in apoptosis in neuronal tissues, promoting cell death through a caspase-dependent mechanism.
Specifications

Specifications

PHD3
Polyclonal
TBS with 0.1% BSA and 0.09% sodium azide
Q91UZ4, Q9H6Z9
EGL9 homolog of C. elegans 3, EGLN3, Factor responsive smooth muscle protein, HIF Prolyl Hydroxylase 3, HIFP4H3, HIFPH3, P4H3, Prolyl Hydroxylase Doma
Rabbit
IgG
100 μl
4° C
Primary
112399, 112407
Western Blot
Unconjugated
EGLN3
liquid
Egln3
Between 50 and 100
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit