Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHD4/HIF Prolyl Hydroxylase 4 Antibody (SR1816), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | PHD4/HIF Prolyl Hydroxylase 4 |
---|---|
Clone | SR1816 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation |
Classification | Monoclonal |
Description
PHD4/HIF Prolyl Hydroxylase 4 Monoclonal antibody specifically detects PHD4/HIF Prolyl Hydroxylase 4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunoprecipitationSpecifications
PHD4/HIF Prolyl Hydroxylase 4 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 1.14.11.-, EGLN4, HIFPH4, HIF-PH4, HIF-prolyl hydroxylase 4, HPH-4, hypoxia-inducible factor prolyl 4-hydroxylase, Hypoxia-inducible factor prolyl hydroxylase 4, P4H with transmembrane domain, P4H-TMFLJ20262, PH-4, PH4 hypoxia inducible factor prolyl 4 hydroxylase, PHD4, proline 4-hydroxylase, prolyl 4-hydroxylase, transmembrane (endoplasmic reticulum), Prolyl hydroxlase domain-containing 4, transmembrane prolyl 4-hydroxylase | |
A synthesized peptide derived from human PHD4/HIF Prolyl Hydroxylase 4 (Uniprot #: Q9NXG6) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1816 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cancer, Hypoxia, Lipid and Metabolism | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
54681 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title