Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phenacetin melting point standard Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
1 g
Description
Pharmaceutical (small molecule)

Specifications
Specifications
CAS | 62-44-2 |
Physical Form | Neat |
Linear Formula | CH3 CONHC6H4 OC2H5 |
MDL Number | MFCD00009094 |
Melting Point | ∼135°C; 133°C to 136°C |
Synonym | Phenacetin; 1-Acetyl-p-phenetidin; 4 ′-Ethoxyacetanilide; N-(4-Ethoxyphenyl)acetamide; p-Acetophenetidide; Acetophenetidin |
Type | Certified Reference Material |
Quantity | 1 g |
Formula Weight | 179.22 |
Grade | Certified Reference Material |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction