Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenanthrene 90%, ACROS Organics™

Click to view available options
Quantity:
1kg
85-01-8
C14H10
178.234
MFCD00001168
YNPNZTXNASCQKK-UHFFFAOYSA-N
phenanthren,phenanthrin,phenanthracene,ravatite,phenantrin,phenanthren german,phenanthrene, pure,unii-448j8e5bst,ccris 1233,chembl46730
995
CHEBI:28851
phenanthrene
C1=CC=C2C(=C1)C=CC3=CC=CC=C32
85-01-8
C14H10
178.234
MFCD00001168
YNPNZTXNASCQKK-UHFFFAOYSA-N
phenanthren,phenanthrin,phenanthracene,ravatite,phenantrin,phenanthren german,phenanthrene, pure,unii-448j8e5bst,ccris 1233,chembl46730
995
CHEBI:28851
phenanthrene
C1=CC=C2C(=C1)C=CC3=CC=CC=C32
Color White to Brown
Quantity 1kg
Density 1.063g/mL
Infrared Spectrum Authentic
Melting Point 95°C to 101°C
Assay Percent Range 90%
Beilstein 05, 667
Merck Index 15, 7326
Formula Weight 178.23
Physical Form Crystalline Powder or Flakes
Percent Purity ≥88%
Chemical Name or Material Phenanthrene, 90%
Show More Show Less

GHS H Statement:
Suspected of causing cancer.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.

GHS P Statement:
Use personal protective equipment as required.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning