Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenazine methosulfate, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
25g
Packaging:
Glass bottle
299-11-6
C13H11N2·CH3O4S
306.33
MFCD00011923
RXGJTUSBYWCRBK-UHFFFAOYSA-M
phenazine methosulfate,5-methylphenazin-5-ium methyl sulfate,5-methylphenazinium methyl sulfate,n-methylphenazonium methosulfate,phenazine methosulphate,methylphenazonium methosulfate,5-methylphenazine methylsulfate,n-methylphenazinium methosulfate,n-methylphenazonium methosulphate,phenazinium, 5-methyl-, methyl sulfate
9285
CHEBI:8055
5-methylphenazin-5-ium;methyl sulfate
C[N+]1=C2C=CC=CC2=NC3=CC=CC=C31.COS(=O)(=O)[O-]
299-11-6
C13H11N2·CH3O4S
306.33
MFCD00011923
RXGJTUSBYWCRBK-UHFFFAOYSA-M
phenazine methosulfate,5-methylphenazin-5-ium methyl sulfate,5-methylphenazinium methyl sulfate,n-methylphenazonium methosulfate,phenazine methosulphate,methylphenazonium methosulfate,5-methylphenazine methylsulfate,n-methylphenazinium methosulfate,n-methylphenazonium methosulphate,phenazinium, 5-methyl-, methyl sulfate
9285
CHEBI:8055
5-methylphenazin-5-ium;methyl sulfate
C[N+]1=C2C=CC=CC2=NC3=CC=CC=C31.COS(=O)(=O)[O-]
Melting Point 158°C to 160°C
Packaging Glass bottle
Color Brown to Yellow or Khaki
Quantity 1g
Infrared Spectrum Authentic
Assay Percent Range 98%
Merck Index 15, 6182
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol and ethanol,insoluble in benzene and diethylether
Formula Weight 306.33
Physical Form Crystalline Powder
Chemical Name or Material Phenazine methosulfate, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Suspected of causing genetic defects.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

MUTAGEN

EINECSNumber : 206-072-1

RTECSNumber : SG1645000

TSCA : TSCA