Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenethyl alcohol, 99%, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
250mL
1L
2.5L
10L
Packaging:
Glass bottle
Plastic Jerrycan
60-12-8
C8H10O
122.167
MFCD00002886
WRMNZCZEMHIOCP-UHFFFAOYSA-N
phenethyl alcohol,benzeneethanol,phenylethyl alcohol,benzyl carbinol,phenylethanol,phenethanol,2-phenylethyl alcohol,2-phenyl-ethanol,beta-phenylethanol,2-phenylethan-1-ol
6054
CHEBI:49000
2-phenylethanol
C1=CC=C(C=C1)CCO
60-12-8
C8H10O
122.167
MFCD00002886
WRMNZCZEMHIOCP-UHFFFAOYSA-N
phenethyl alcohol,benzeneethanol,phenylethyl alcohol,benzyl carbinol,phenylethanol,phenethanol,2-phenylethyl alcohol,2-phenyl-ethanol,beta-phenylethanol,2-phenylethan-1-ol
6054
CHEBI:49000
2-phenylethanol
C1=CC=C(C=C1)CCO
Refractive Index 1.5300 to 1.5350
Flash Point 102°C
Viscosity 7.58 mPa.s (25°C)
Specific Gravity 1.02
Boiling Point 219.0°C
Melting Point -27.0°C
Packaging Plastic Jerrycan
Infrared Spectrum Authentic
Quantity 10L
Density 1.0200g/mL
Color Colorless
Assay Percent Range 99%
Linear Formula C6H5CH2CH2OH
Beilstein 06, 478
Merck Index 15, 7338
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: miscible with alcohol and ether
Formula Weight 122.17
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Phenethyl alcohol
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

Danger