Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenethyl isocyanate, 99%, Acros Organics

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
1943-82-4
C9H9NO
147.18
MFCD00037044
HACRKYQRZABURO-UHFFFAOYSA-N
phenethyl isocyanate,2-isocyanatoethyl benzene,phenylethyl isocyanate,2-phenylethyl isocyanate,2-phenylethylisocyanate,phenylethylisocyanate,phenethylisocyanate,benzene, 2-isocyanatoethyl,2-isocyanato-ethyl-benzene,2-phenethyl isocyanate
160602
CHEBI:60067
2-isocyanatoethylbenzene
C1=CC=C(C=C1)CCN=C=O
1943-82-4
C9H9NO
147.18
MFCD00037044
HACRKYQRZABURO-UHFFFAOYSA-N
phenethyl isocyanate,2-isocyanatoethyl benzene,phenylethyl isocyanate,2-phenylethyl isocyanate,2-phenylethylisocyanate,phenylethylisocyanate,phenethylisocyanate,benzene, 2-isocyanatoethyl,2-isocyanato-ethyl-benzene,2-phenethyl isocyanate
160602
CHEBI:60067
2-isocyanatoethylbenzene
C1=CC=C(C=C1)CCN=C=O
Boiling Point 210°C
Flash Point 99°C
Specific Gravity 1.063
Refractive Index 1.5210 to 1.523
Quantity 25mL
Infrared Spectrum Authentic
Density 1.0630g/mL
Packaging Glass bottle
Color Colorless to Yellow
Assay Percent Range 99%
Linear Formula C6H5(CH2)2NCO
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in toluene
Formula Weight 147.18
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Phenethyl isocyanate
Show More Show Less

GHS H Statement
May cause allergy or asthma symptoms or breathing difficulties if inhaled. Harmful if swallowed. Toxic if inhaled. Toxic to aquatic life with long lasting effects. Causes severe skin burns and eye damage. Causes serious eye damage. May cause an allergic skin reaction.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. Wear eye protection/face protection. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

Danger