Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenethylamine, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
250g
1kg
5kg
Packaging:
Glass bottle
Plastic drum
64-04-0
C8H11N
121.18
MFCD00008184
BHHGXPLMPWCGHP-UHFFFAOYSA-N
phenethylamine,2-phenylethylamine,benzeneethanamine,phenylethylamine,2-phenethylamine,beta-phenylethylamine,beta-phenethylamine,1-amino-2-phenylethane,2-aminoethyl benzene,beta-aminoethylbenzene
1001
CHEBI:18397
2-phenylethanamine
C1=CC=C(C=C1)CCN
64-04-0
C8H11N
121.18
MFCD00008184
BHHGXPLMPWCGHP-UHFFFAOYSA-N
phenethylamine,2-phenylethylamine,benzeneethanamine,phenylethylamine,2-phenethylamine,beta-phenylethylamine,beta-phenethylamine,1-amino-2-phenylethane,2-aminoethyl benzene,beta-aminoethylbenzene
1001
CHEBI:18397
2-phenylethanamine
C1=CC=C(C=C1)CCN
Viscosity 2.4 mm2/s (23°C)
Water 0.5% max.
Flash Point 81°C
Refractive Index 1.5320 to 1.5340
Packaging Plastic drum
Infrared Spectrum Authentic
Specific Gravity 0.985
Quantity 5kg
Melting Point -60.0°C
Color Colorless to Yellow
Boiling Point 200.0°C to 202.0°C
Density 0.9850g/mL
Assay Percent Range 98.5% min. (GC)
Linear Formula C6H5CH2CH2NH2
Beilstein 12, 1096
Fieser 09,363
Merck Index 15, 7339
Solubility Information Solubility in water: 63g/L (25°C). Other solubilities: soluble in ethanol, diethyl ether
Formula Weight 121.18
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Phenethylamine
Show More Show Less

GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 200-574-4