Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Phenetole, 98+%

Click to view available options
Quantity:
100mL
500mL
2500mL
103-73-1
C8H10O
122.167
MFCD00009090
DLRJIFUOBPOJNS-UHFFFAOYSA-N
phenetole,benzene, ethoxy,ethyl phenyl ether,phenetol,phenyl ethyl ether,phenoxyethane,a phenoxyethane,ether, ethyl phenyl,unii-rb8lu2c57f
7674
CHEBI:67129
ethoxybenzene
CCOC1=CC=CC=C1
103-73-1
C8H10O
122.167
MFCD00009090
DLRJIFUOBPOJNS-UHFFFAOYSA-N
phenetole,benzene, ethoxy,ethyl phenyl ether,phenetol,phenyl ethyl ether,phenoxyethane,a phenoxyethane,ether, ethyl phenyl,unii-rb8lu2c57f
7674
CHEBI:67129
ethoxybenzene
CCOC1=CC=CC=C1
Refractive Index 1.507
Density 0.967
Quantity 500mL
Odor Aromatic
Boiling Point 169°C to 170°C
Flash Point 57°C (134°F)
Melting Point -30°C
UN Number UN3271
Beilstein 636270
Merck Index 14,7229
Formula Weight 122.17
Percent Purity ≥98%
Chemical Name or Material Phenetole
Show More Show Less

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.

EINECSNumber : 203-139-7

RTECSNumber : SI7700000

TSCA : Yes

Recommended Storage : Ambient temperatures