Learn More
Medchemexpress LLC Phenol, 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)- | 269409-70-3 | 220.08 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW002879100G
Encompass_Preferred
4-Hydroxyphenylboronic acid pinacol ester is a ROS-responsive component. It can structurally modify hyaluronic acid to prepare ROS-responsive intelligent nanocarriers and compound delivery systems. It endows hyaluronic acid-based nanocarriers with ROS responsiveness, enabling them to rapidly release encapsulated Curcumin in the presence of reactive oxygen species. This product is applicable to research related to periodontitis and can also be used for the synthesis of boronic acid-modified resins.
  • ROS-responsive component
  • Can structurally modify hyaluronic acid to prepare ROS-responsive intelligent nanocarriers and compound delivery systems
  • Endows hyaluronic acid-based nanocarriers with ROS responsiveness, allowing for rapid release of encapsulated curcumin in the presence of reactive oxygen species
  • Research related to periodontitis
  • Synthesis of boronic acid-modified resins
Catalog No. 50-004-45360