Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol, 99+%, for biochemistry, loose crystals, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic Drum
108-95-2
C6H6O
94.113
MFCD00002143
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
carbolic acid,hydroxybenzene,phenic acid,phenylic acid,oxybenzene,benzenol,phenyl hydrate,monophenol,phenyl hydroxide,phenylic alcohol
996
CHEBI:15882
phenol
C1=CC=C(C=C1)O
108-95-2
C6H6O
94.113
MFCD00002143
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
carbolic acid,hydroxybenzene,phenic acid,phenylic acid,oxybenzene,benzenol,phenyl hydrate,monophenol,phenyl hydroxide,phenylic alcohol
996
CHEBI:15882
phenol
C1=CC=C(C=C1)O
Water 0.5% max.
Residue after Evaporation 0.05% max.
Color Colorless to White
Infrared Spectrum Authentic
Density 1.0700g/mL
Quantity 2.5kg
Packaging Plastic Drum
Assay Percent Range 99+%
Linear Formula C6H5OH
Beilstein 06, 110
Fieser 01,828; 13,166; 14,88
Merck Index 15, 7354
Formula Weight 94.11
Physical Form Loose Crystals
Percent Purity ≥99.0%
Grade Biochemical
Chemical Name or Material Phenol, 99+%
Show More Show Less

GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing genetic defects.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 203-632-7

RTECSNumber : SJ3325000

TSCA : TSCA