Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol, 99%, extra pure, ACROS Organics™

Click to view available options
Quantity:
50g
1kg
2.5kg
5kg
Packaging:
Glass bottle
Plastic drum
108-95-2
C6H6O
94.11
MFCD00002143
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
carbolic acid,hydroxybenzene,phenic acid,phenylic acid,oxybenzene,benzenol,phenyl hydrate,monophenol,phenyl hydroxide,phenylic alcohol
996
CHEBI:15882
phenol
C1=CC=C(C=C1)O
108-95-2
C6H6O
94.11
MFCD00002143
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
carbolic acid,hydroxybenzene,phenic acid,phenylic acid,oxybenzene,benzenol,phenyl hydrate,monophenol,phenyl hydroxide,phenylic alcohol
996
CHEBI:15882
phenol
C1=CC=C(C=C1)O
Boiling Point 182.0°C
Flash Point 79°C
Vapor Pressure 0.4mbar at 20°C
Packaging Glass bottle
Specific Gravity 1.07
Water 0.2% max.
Density 1.0700g/mL
Infrared Spectrum Authentic
Melting Point 39.0°C to 42.0°C
Color Colorless
Quantity 1kg
Viscosity 3.437 mPa.s (50°C)
Assay Percent Range 99%
Linear Formula C6H5OH
Beilstein 06, 110
Fieser 01,828; 13,166; 14,88
Merck Index 15, 7354
Solubility Information Solubility in water: 8.4g/L (20°C). Other solubilities: soluble in aq. alkali hydroxides, soluble in alcohol, chloroform, ether, glycerol,, carbon disulfide and fixed oils, 1g/12mL benzene, practically insoluble in petroleum ether
Formula Weight 94.11
Physical Form Crystals or Fused Mass
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material Phenol, 99%
Show More Show Less

GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing genetic defects.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

Recommended Storage : Turns pink on exposure to air