Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol ACS AR Loose Crystals, J.T.Baker™

Click to view available options
Quantity:
500g
12 x 125g
12 x 500g
Packaging:
Glass
Poison Pack
108-95-2
C6H6O
94.113
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
996
CHEBI:15882
phenol
C1=CC=C(C=C1)O
108-95-2
C6H6O
94.113
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
996
CHEBI:15882
phenol
C1=CC=C(C=C1)O
Packaging Glass
Quantity 12 x 500g
Formula Weight 94.11
Physical Form Loose Crystals
Grade ACS
Chemical Name or Material Phenol