Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol:Chloroform Saturated Solution, pH 5.2, MP Biomedicals

This is a ready-to-use saturated solution of phenol and chloroform suitable for RNA purifications. 1:1 solution, pH 5.2

Specifications

Chemical Name or Material Phenol:Chloroform Saturated Solution
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN802515
View Documents
MP Biomedicals, Inc
04802515/DEL
400mL This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Specifications

Specifications

Phenol:Chloroform Saturated Solution
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit