Alfa Aesar™ Phenol-d1, 95% (Isotopic)

Manufacturer: Alfa Aesar™ H5110006

Quantity 5g

 View more versions of this product

Catalog No. AAH5110006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1003-66-3
    Molecular Formula C6H5DO
    Formula Weight 95.12
    MDL Number MFCD01075468
    UN Number UN1671
    Synonym Phenol-4-d1
    Chemical Name or Material Phenol-d1, Isotopic

    Specifications

    Formula Weight 95.12
    Boiling Point 182°C
    Melting Point 40° to 42°C
    Quantity 5g
    Molecular Formula C6H5DO
    MDL Number MFCD01075468
    CAS 1003-66-3
    UN Number UN1671
    Synonym Phenol-4-d1
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H314-H318-H341-H373
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Suspected of causing genetic defects.
    May cause damage to organs through prolonged or repeated exposure.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: PHENOL, SOLID
    Chemical Name or Material Phenol-d1, Isotopic
    Assay 95%

    GHS H Statement
    H300-H310-H330-H314-H318-H341-H373
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Suspected of causing genetic defects.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit