Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol, Fused Crystal, White, Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
100g
500g
1kg
Packaging:
Steel Can
108-95-2
C6H6O
94.113
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
carbolic acid,hydroxybenzene,phenic acid,phenylic acid,oxybenzene,benzenol,phenyl hydrate,monophenol,phenyl hydroxide,phenylic alcohol
996
CHEBI:15882
phenol
C1=CC=C(C=C1)O
108-95-2
C6H6O
94.113
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
carbolic acid,hydroxybenzene,phenic acid,phenylic acid,oxybenzene,benzenol,phenyl hydrate,monophenol,phenyl hydroxide,phenylic alcohol
996
CHEBI:15882
phenol
C1=CC=C(C=C1)O

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Extraction of Nucleic Acids and Molecular Biology Applications
Color White
Quantity 1kg
Flash Point 79.00°C
Packaging Steel Can
Specific Gravity 1L = 1.06kg
Formula Weight 94.11
Physical Form Fused Crystal
Grade Ultrapure Bioreagent
Chemical Name or Material Phenol