Phenol (Fused Crystals/Laboratory), Fisher Chemical

Packaging Glass Bottle
Quantity 500g

 View more versions of this product

Catalog No. A91I-500

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 108-95-2
    Physical Form Solid
    Molecular Formula C6H6O
    Linear Formula C6H5OH
    Formula Weight 94.11
    MDL Number MFCD00002143
    Synonym Carbolic Acid
    Chemical Name or Material Phenol
    Grade Laboratory Grade

    Specifications

    Boiling Point 182°C
    Formula Weight 94.11
    Melting Point 39°C
    pH 4 to 5
    Quantity 500g
    UN Number 1671
    Flash Point 79°C
    DOT Information DOT Class 6.1, : Poison
    Physical Form Solid
    Color Colorless
    Density 3.2
    Viscosity 12.7cP
    Vapor Pressure 0.4mbar at 20°C
    Health Hazard 3 Emergency Overview
    Combustible material. Causes burns by all exposure routes. Danger of serious damage to health by prolonged exposure. Possible risks of irreversible effects. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

    NFPA

    Health:3
    Flammability:2
    Instability:1
    Identification Pass Test
    Packaging Glass Bottle
    Molecular Formula C6H6O
    Linear Formula C6H5OH
    MDL Number MFCD00002143
    CAS 108-95-2
    ChemAlert Storage Symbol White
    Synonym Carbolic Acid
    Additional Information Vapor Pressure: 0.4mbar at 20°C
    Health Hazard 2 DANGER!
    Chemical Name or Material Phenol
    Grade Laboratory Grade
     

    DANGER!

    Emergency Overview
    Combustible material. Causes burns by all exposure routes. Danger of serious damage to health by prolonged exposure. Possible risks of irreversible effects. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

    NFPA

    Health:3
    Flammability:2
    Instability:1

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit