Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol, Liquefied, Contains about 20ppm Citric Acid as a Preservative, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Manufacturer:  J.T.Baker 285901

Packaging Narrow Mouth Amber Glass Bottle
Quantity 500mL

 View more versions of this product

Catalog No. 02-003-676

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

108-95-2
94.11
BAKER ANALYZED™ Reagent
C6H5OH
Phenol, Liquefied, Contains about 20ppm Citric Acid as a Preservative

Specifications

500mL
Narrow Mouth Amber Glass Bottle
94.11
79.00°C
BAKER ANALYZED™ Reagent
108-95-2
C6H5OH
1L = 1.06kg
Phenol, Liquefied, Contains about 20ppm Citric Acid as a Preservative
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit