Phenol, Liquefied, Contains about 20ppm Citric Acid as a Preservative, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Manufacturer: J.T.Baker 285904

Packaging Narrow Mouth Amber Glass Bottle
Quantity 150mL

 View more versions of this product

Catalog No. 02-003-674

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 108-95-2
    Molecular Formula C6H5OH
    Formula Weight 94.11
    Chemical Name or Material Phenol, Liquefied, Contains about 20ppm Citric Acid as a Preservative
    Grade BAKER ANALYZED™ Reagent

    Specifications

    Formula Weight 94.11
    Molecular Formula C6H5OH
    CAS 108-95-2
    Quantity 150mL
    Packaging Narrow Mouth Amber Glass Bottle
    Specific Gravity 1L = 1.06kg
    Flash Point 79.00°C
    Chemical Name or Material Phenol, Liquefied, Contains about 20ppm Citric Acid as a Preservative
    Grade BAKER ANALYZED™ Reagent

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit