Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol, Liquefied, Contains about 20ppm Citric Acid as a Preservative, Multi-Compendial, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle

Chemical Identifiers

108-95-2
C6H6O
94.113
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
996
CHEBI:15882
Quantity 500mL
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.06kg
SMILES C1=CC=C(C=C1)O
IUPAC Name phenol
Formula Weight 94.11
Flash Point 79.00°C
Chemical Name or Material Phenol, Liquefied, Contains about 20ppm Citric Acid as a Preservative
Grade Multi-Compendial