Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol, Loose Crystal, Technical, Spectrum™

Click to view available options
Quantity:
25 g
1 kg
2.5 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
108-95-2
108-95-2
Packaging Amber Glass Bottle
Quantity 1 kg
CAS Max % 1
Grade Technical

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.