Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol, Molecular Biology Grade, Calbiochem™, EMD Millipore

Manufacturer:  Millipore Sigma 516724100GM

Quantity 100g

 View more versions of this product

Catalog No. 51-672-4100GM

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

108-95-2
C6H6O
Colorless
40.8°C
Solid
94.1
UN 1671
Phenol

Specifications

100g
Packaged under inert gas
Solid
94.1
Water: 84g/L at 20°C soluble
1.06g/cm3 (20°C)
181.8°C
5 (50g/L at 20°C)
595°C (1103°F)
≥99.0%
108-95-2
Characteristic
C6H6O
UN 1671
0.2 hPa (20°C)
Colorless
40.8°C
81°C (178°F)
Phenol
GC Grade, Molecular biology grade

Safety and Handling

H314: Causes severe skin burns and eye damage.

H341: Suspected of causing genetic defects.

H301-H311-H331: Toxic if swallowed. In contact with skin or if inhaled

DOTInformation : DOT Class: 6.1, Packaging Group: II

RTECSNumber : SJ3325000

Recommended Storage : -20°C (-4°F), Tightly closed. Dry. Keep in a well-ventilated place. Keep locked up or in an area accessible only to qualified or authorized person.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit