Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Phenol, Ready-to-Use, buffer saturated solution pH 4.5

Click to view available options
Quantity:
100mL
400mL
108-95-2
C6H6O
94.113
MFCD00002143
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
carbolic acid,hydroxybenzene,phenic acid,phenylic acid,oxybenzene,benzenol,phenyl hydrate,monophenol,phenyl hydroxide,phenylic alcohol
996
CHEBI:15882
phenol
C1=CC=C(C=C1)O
108-95-2
C6H6O
94.113
MFCD00002143
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
carbolic acid,hydroxybenzene,phenic acid,phenylic acid,oxybenzene,benzenol,phenyl hydrate,monophenol,phenyl hydroxide,phenylic alcohol
996
CHEBI:15882
phenol
C1=CC=C(C=C1)O
UN Number UN2821
Beilstein 6428463
Density 1.06g/mL
Formula Weight 94.11
Physical Form Liquid
Color Undesignated
pH 4.5
Quantity 100mL
Sensitivity Light Sensitive
Chemical Name or Material Phenol, Ready-to-Use, buffer saturated solution

GHS H Statement
H311-H331-H314-H341-H373-H227-H302
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: PHENOL SOLUTION

TSCA : TSCA