Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol Red Indicator, 0.04% (w/v), Ricca Chemical
SDP

Catalog No. p-2916771
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle

Specifications

Chemical Name or Material Phenol Red Indicator
CAS 7732-18-5, 34487-61-1
Percent Purity 0.04% W/V
Grade Indicator
Concentration or Composition (by Analyte or Components) 0.04% (w/v) Aqueous Solution
Packaging Natural Poly Bottle
Color Orange/Red
pH 6 to 7.3
Quantity 120 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.