Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phenol Resin 100-200 Mesh 1percent , Advanced ChemTech
Click to view available options
Quantity:
1 g
5 g
25 g
Specifications
Specifications
Color | White/Off-White |
Quantity | 5 g |
Grade | Biochemical |
Chemical Name or Material | Phenol Resin 100-200 Mesh 1% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction