Learn More
Fisherbrand™ Phenolic Standing Height Cabinet
Fisherbrand™ Phenolic Casework is designed to rugged use in wet areas where corrosion or rust are of concern.
Description
- Rust, rot, mold resistant construction.
- Cabinets will not absorb odors, are easy to clean and are virtually maintenance free.
- Standard cabinet construction is from 0.5 in. thick chemically resistant phenolic panels in a full overlay style.
- All colors have a solid black core
Specifications
2 years | |
1 | |
34.875 in. | |
2 | |
Platinum | |
55.9 cm | |
Phenolic |
121.9 cm | |
48 in. | |
2 | |
Standing Height | |
22 in. | |
88.6 cm |